missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ ProCare Lab 10 MA - 5 L

Product Code. 15430425 Shop All Miele Products
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15430425 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430425 Supplier Miele Professional™ Supplier No. 10265670
  • ONLINE EXCLUSIVE PRICE
    £223.66
    £58.00 / 5 L
    Save £165.66
    74% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid detergent, mildly alkaline, 5 L. For optimum reprocessing of laboratory glassware and utensils.

  • Free from surfactants and oxidants
  • Specially developed for Miele Professional
  • Suitable for all types of materials
  • pH value 11.6 to 11.8
TRUSTED_SUSTAINABILITY

Specifications

Product Type Liquid Detergent
pH 11.6 to 11.8
Description Liquid detergent, mildly alkaline
For Use With (Equipment) For optimum reprocessing of laboratory glassware and utensils
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • Miele ProCare Lab 10 MA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.