Description
- Universal alkaline cleaning agent
- Free from oxidants and surfactants
- Specially developed for Miele Professional washer-disinfectors
- Suitable for stainless steel, laboratory glassware, ceramics and plastics
- Effective cleaning for light to medium soiling
Specifications
Specifications
| Product Type | Cleaning Agent |
| Color-coded | Yes |
| Form | Liquid |
| Scent | Unscented |
| Sterility | Non-sterile |
| Quantity | 10 L |