missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Detergent

Product Code. 15506935 Shop All Miele Products
Change view
Click to view available options
pH:
12.5 to 12.7
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
15506935 12.5 to 12.7 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15506935 Supplier Miele Professional™ Supplier No. 10265620
  • ONLINE EXCLUSIVE PRICE
    £65.99
    £55.00 / 5 L
    Save £10.99
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid detergent ProCare Lab 10 AP, alkaline, 5 L

  • Free from surfactants, phosphorous compounds and oxidants
  • Specially developed for Miele Professional
  • Suitable for all types of materials
  • pH value from 12.5 to 12.7
  • For use with laboratory glassware washers
  • Excellent for the removal of low-viscosity oils, inorganic and organic soiling, and stubborn or dried-on residues

TRUSTED_SUSTAINABILITY

Specifications

Form Concentrated Liquid
pH 12.5 to 12.7
Composition Polycarboxylate Alkali hydroxide
Description ProCare Lab 10 AP - 5 L
For Use With (Application) Cleaning of lab utensils
For Use With (Equipment) Lab washers
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • Miele ProCare Lab 10 AP
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.