missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Procare Hand Cream

Product Code. 11877462
Change view
Click to view available options
Quantity:
4 Pk.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11877462 4 Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11877462 Supplier Day Impex™ Supplier No. 339025
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Destroy a broad specturm of organisms with a hand cream specially formulated for long-lasting skin care protection. Day Impex™ ProCARE Hand Cream is a non-greasy professional barrier hand cream that relieves and protects hands from chapping and irritation.

  • Particularly effective in reducing the irritation caused by the constant wearing of rubber gloves
  • Combining lanolin to moisturise the skin and silicone to seal in that moisture
  • Absorbs rapidly for a non-greasy feel and instant protection.
  • Thick cream formulation so it can be used sparingly.
  • A single 85 g tube will last several weeks
  • In biohazardous situations, we recommend a dedicated tube for each user to reduce tbe risk of cross-contamination

Specifications

Product Type Procare Hand Cream
Color Off-white
Form Cream
Composition Cetyl Alcohol Dimethicone, Sorbitol Glyceryl Stearate, Sodium Lauryl Sulphate PEG-60 Lanolin, Nonoxynol-9 (permissible minimum) Propylene Glycol, Diazolidinyl Urea, Methylparaben, Propylparaben Water, Kathon CG
For Use With (Application) Laboratory and Healthcare Personnel
Packaging Pump Top Dispenser
Quantity 4 Pk.
Volume (Metric) 450 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.