missing translation for 'onlineSavingsMsg'
Learn More

Procarbazine Hydrochloride, TRC

Product Code. 30422535
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422535 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422535 Supplier Toronto Research Chemicals Supplier No. TRCP755400250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Procarbazine hydrochloride
CAS 366-70-1
Molecular Formula C12 H19 N3 O . Cl H
Quantity 250 mg
Synonym Benzamide','N-(1-methylethyl)-4-[(2-methylhydrazinyl)methyl]-','hydrochloride (1:1), Benzamide','N-(1-methylethyl)-4-[(2-methylhydrazino)methyl]-','monohydrochloride (9CI), p-Toluamide','N-isopropyl-α-(2-methylhydrazino)-,monohydrochloride (8CI), 1-Methyl-2-[p-(isopropylcarbamoyl)benzyl]hydrazine hydrochloride, 2-[-Isopropylcarbamoyl)benzyl]-1-methylhydrazine hydrochloride, IBZ, Ibenzmethyzin hydrochloride, Ibenzmethyzine hydrochloride, MBH, MIH hydrochloride, Matulane, N-Isopropyl-p-(2-methylhydrazinomethyl)benzamide hydrochloride, N-Isopropyl-α-(2-methylhydrazino)-p-toluamide hydrochloride, NCI-C 01810, NSC 77213, Nathulane, Natulan, Natulan hydrochloride, Natulanar, PCB hydrochloride, Procarbazine hydrochloride, Ro 4 6467/1, Ro 4-6467
InChI Formula InChI=1S/C12H19N3O.ClH/c1-9(2)15-12(16)11-6-4-10(5-7-11)8-14-13-3;/h4-7,9,13-14H,8H2,1-3H3,(H,15,16);1H
SMILES Cl.CNNCc1ccc(cc1)C(=O)NC(C)C
IUPAC Name 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
Molecular Weight (g/mol) 257.76
Formula Weight 257.1295
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.