Learn More
Invitrogen™ Procalcitonin Human ELISA Kit
Sandwich ELISA Kit
£554.00 - £2320.00
Specifications
Accession Number | P01258 |
---|---|
Assay Sensitivity | 30 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465093
|
Invitrogen™
EHPCT |
96 Tests |
£554.00
1 set |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
17195529
|
Invitrogen™
EHPCTX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Human Procalcitonin ELISA quantitates Hu Procalcitonin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Procalcitonin. Principle of the method The Human Procalcitonin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
P01258 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
CALC1,CGRP,CGRP-I,CGRP1,CT,KC,PCT | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
30 pg/mL | |
Biotin | |
GCPR | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
796 | |
<12% | |
HRP | |
Neurobiology | |
CALC1, CGRP, CGRP-I, CGRP1, CT, KC | |
1 hr. 20 min. |