missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00233-100ul
This item is not returnable.
View return policy
Description
Procalcitonin Polyclonal antibody specifically detects Procalcitonin in Human samples. It is validated for ELISA
Specifications
| Procalcitonin | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Produced in rabbits immunized with purified, recombinant Human Procalcitonin (Accession#: P01258-1; Ala26-Asn141) | |
| 100 μg | |
| Cancer, Cellular Markers, Neuroscience | |
| 796 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction