missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (6f10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-13044
This item is not returnable.
View return policy
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Procalcitonin | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CALCA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
| 0.2 mg | |
| Cancer | |
| 796 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 6f10 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human PCT | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction