missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (6f10), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-13044AF350
This item is not returnable.
View return policy
Description
Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA
Specifications
| Procalcitonin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
| 0.2 mL | |
| Cancer, Cellular Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 796 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 6f10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction