missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (38F11), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-79065B
This item is not returnable.
View return policy
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| Procalcitonin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CALCA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
| 0.2 mL | |
| Cancer | |
| 796 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 38F11 | |
| Western Blot, ELISA, Sandwich ELISA | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human PCT | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction