missing translation for 'onlineSavingsMsg'
Learn More

Procainamide hydrochloride, 99%

Product Code. 10167743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167743 100 g Glass bottle 100g
10733211 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167743 Supplier Thermo Scientific Acros Supplier No. 207301000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 614-39-1 | C13H21N3O·HCl | 271.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-39-1
Molecular Formula C13H21N3O·HCl
Molecular Weight (g/mol) 271.78
MDL Number MFCD00012998
InChI Key ABTXGJFUQRCPNH-UHFFFAOYSA-N
Synonym procainamide hydrochloride, procainamide hcl, pronestyl, procanbid, procapan, procan, procainhydrochlorid, procan sr, procainii chloridum, promide hydrochloride
PubChem CID 66068
ChEBI CHEBI:8429
IUPAC Name 4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
SMILES CCN(CC)CCNC(=O)C1=CC=C(C=C1)N.Cl

Specifications

Melting Point 166°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl
Quantity 100 g
Beilstein 14, III, 1077
Merck Index 15, 7874
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in chloroform,very sparingly soluble in benzene and ether
Formula Weight 271.78
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Procainamide hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Procainamide hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.