missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Probucol, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
Description
Probucol inhibits atherogenesis in genetically hypercholesterolemic rabbits (Watanabe) and attenuates ischemia/reperfusion-induced cardiomyocyte apoptosis.
Specifications
Specifications
| CAS | 23288-49-5 |
| Melting Point | 124°C to 126°C (literature) |
| Molecular Formula | C31H48O2S2 |
| Quantity | 10 g |
| Synonym | probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum |
| Solubility Information | Soluble in ethanol (50mg/mL, clear and colorless solution). |
| InChI Key | FYPMFJGVHOHGLL-UHFFFAOYSA-N |
| SMILES | CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C |
| IUPAC Name | 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol |
| Molecular Weight (g/mol) | 516.843 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction