missing translation for 'onlineSavingsMsg'
Learn More

Probenecid, TRC

Product Code. 30135310
Change view
Click to view available options
Quantity:
50 mg
5 g
100 g
Unit Size:
100g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135310 100 g 100g
30132429 5 g 5g
17943175 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135310 Supplier Toronto Research Chemicals Supplier No. TRCP755000100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13 H19 N O4 S
Molecular Weight (g/mol) 285.36
Synonym Probenecid, Benzoic acid, 4-[(dipropylamino)sulfonyl]-, Benzoic acid, p-(dipropylsulfamoyl)- (8CI), 4-[(Dipropylamino)sulfonyl]benzoic acid, 4-(Dipropylsulfamoyl)benzoic acid, 4-(N,N-Dipropylaminosulfonyl)benzoic acid, 4-(N,N-Dipropylsulfamoyl)benzoic acid, Apurina, Benecid, Benemid, Benuryl, NSC 18786, Probecid, Proben, Probenecid, Probenecid acid, Prolongine, Synergid R, Tubophan, Uricosid, p-(Dipropylsulfamoyl)benzoic acid, p-(Dipropylsulfamyl)benzoic acid
IUPAC Name 4-(dipropylsulfamoyl)benzoic acid
SMILES CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C13H19NO4S/c1-3-9-14(10-4-2)19(17,18)12-7-5-11(6-8-12)13(15)16/h5-8H,3-4,9-10H2,1-2H3,(H,15,16)
Formula Weight 285.1035
Chemical Name or Material Probenecid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.