missing translation for 'onlineSavingsMsg'
Learn More

Probenecid, MedChemExpress

Product Code. 16440988
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16440988 5 g 5g
16402229 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16440988 Supplier Medchem Express Supplier No. HYB0545
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Probenecid is a potent and selective agonist of transient receptor potential vanilloid 2 (TRPV2) channels. Probenecid also inhibits pannexin 1 channels.

Probenecid is a potent and selective agonist of transient receptor potential vanilloid 2 (TRPV2) channels. Probenecid also inhibits pannexin 1 channels.

Specifications

Chemical Name or Material Probenecid
CAS 57-66-9
Color White
Purity Grade Notes Research
Molecular Formula C13H19NO4S
Quantity 5 g
Solubility Information DMSO : ≥ 100 mg/mL (350.43 mM) ∣0.5 M NaOH : 100 mg/mL (350.43 mM; ultrasonic and adjust pH to 12 with NaOH)
SMILES O=C(O)C1=CC=C(S(=O)(N(CCC)CCC)=O)C=C1
Molecular Weight (g/mol) 285.36
Formula Weight 285.36
Percent Purity 99.37%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.