missing translation for 'onlineSavingsMsg'
Learn More

Probenecid-d14, TRC

Product Code. 30412815
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412815 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412815 Supplier Toronto Research Chemicals Supplier No. TRCP75500210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Probenecid-d14
CAS 1189657-87-1
Purity Grade Notes HPLC
Molecular Formula C13 D14 H5 N O4 S
Quantity 10 mg
Synonym Benzoic acid','4-[[di(propyl-1,1,2,2,3,3,3-d7)amino]sulfonyl]- (ACI), 4-[[Di(propyl-1,1,2,2,3,3,3-d7)amino]sulfonyl]benzoic acid (ACI), 4-[Bis(1,1,2,2,3,3,3-heptadeuteriopropyl)sulfamoyl]benzoic acid, Probenecid-d14, Probenecid-D14 (N,N-dipropyl-D14), Probenecid-d14 (di-n-propyl-d14)
InChI Formula InChI=1S/C13H19NO4S/c1-3-9-14(10-4-2)19(17,18)12-7-5-11(6-8-12)13(15)16/h5-8H,3-4,9-10H2,1-2H3,(H,15,16)/i1D3,2D3,3D2,4D2,9D2,10D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N(C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])S(=O)(=O)c1ccc(cc1)C(=O)O
IUPAC Name 4-[bis(1,1,2,2,3,3,3-heptadeuteriopropyl)sulfamoyl]benzoic acid
Molecular Weight (g/mol) 299.445
Formula Weight 299.191
Percent Purity >95
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.