missing translation for 'onlineSavingsMsg'
Learn More

Probenecid Acyl Beta-D-Glucuronide >90%, TRC

Product Code. 30009983
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009983 1 mg 1mg
30008072 5 mg 5mg
30006921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009983 Supplier Toronto Research Chemicals Supplier No. TRCP7550101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34017-15-7
Molecular Formula C19 H27 N O10 S
Molecular Weight (g/mol) 461.48
Synonym Probenecid Acyl-B-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-(dipropylsulfamoyl)benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H27NO10S/c1-3-9-20(10-4-2)31(27,28)12-7-5-11(6-8-12)18(26)30-19-15(23)13(21)14(22)16(29-19)17(24)25/h5-8,13-16,19,21-23H,3-4,9-10H2,1-2H3,(H,24,25)/t13-,14-,15+,16-,19-/m0/s1
Formula Weight 461.1356
Chemical Name or Material Probenecid Acyl beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.