missing translation for 'onlineSavingsMsg'
Learn More

Probenazole, TRC

Product Code. 17947174
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947174 100 mg 100mg
17959654 500 mg 500mg
17998414 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947174 Supplier Toronto Research Chemicals Supplier No. TRCP754085100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27605-76-1
Molecular Formula C10 H9 N O3 S
Molecular Weight (g/mol) 223.25
Synonym 1,2-Benzisothiazole, 3-(2-propenyloxy)-, 1,1-dioxide (9CI), 1,2-Benzisothiazole, 3-(allyloxy)-, 1,1-dioxide (8CI), 3-Allyloxy-1,2-benzisothiazol-1,1-dioxide, 3-Allyloxy-1,2-benzisothiazole 1,1-dioxide, 3-Allyloxy-1,2-benzisothiazole-1,1-oxide, Oryzaemate, Oryzemate, PO 20, PO 20 (pharmaceutical), Probenazole, 1,2-Benzisothiazole, 3-(2-propen-1-yloxy)-, 1,1-dioxide
IUPAC Name 3-prop-2-enoxy-1,2-benzothiazole 1,1-dioxide
SMILES C=CCOC1=NS(=O)(=O)c2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H9NO3S/c1-2-7-14-10-8-5-3-4-6-9(8)15(12,13)11-10/h2-6H,1,7H2
Formula Weight 223.0303
Chemical Name or Material Probenazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.