missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proapoptotic Caspase Adaptor Protein Rabbit anti-Human, Clone: 55, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90321-50UL
This item is not returnable.
View return policy
Description
Proapoptotic Caspase Adaptor Protein Monoclonal specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for ELISA.
Specifications
| Proapoptotic Caspase Adaptor Protein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 55 | |
| ELISA 1:5000-1:10000 | |
| caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein | |
| Recombinant Human Proapoptotic Caspase Adaptor Protein | |
| 50 μg | |
| Apoptosis, Caspases | |
| 51237 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction