missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proapoptotic Caspase Adaptor Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97098-100ul
This item is not returnable.
View return policy
Description
Proapoptotic Caspase Adaptor Protein Polyclonal antibody specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for ELISA
Specifications
| Proapoptotic Caspase Adaptor Protein | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein | |
| Produced in rabbits immunized with purified, recombinant Human Proapoptotic Caspase Adaptor Protein (Uniprot#: Q8WU39-1; Met 1- Thr 185) | |
| 100 μg | |
| Apoptosis, Caspases | |
| 51237 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction