missing translation for 'onlineSavingsMsg'
Learn More

Proapoptotic Caspase Adaptor Protein Antibody (055), FITC, Novus Biologicals™

Product Code. 18340880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340880 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340880 Supplier Novus Biologicals Supplier No. NBP290321F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Proapoptotic Caspase Adaptor Protein Monoclonal specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Proapoptotic Caspase Adaptor Protein
Applications ELISA
Classification Monoclonal
Clone 55
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Proapoptotic Caspase Adaptor Protein (Q8WU39-1; Met 1- Thr 185).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 51237
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.