missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proapoptotic Caspase Adaptor Protein Antibody (022), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90320UV
This item is not returnable.
View return policy
Description
Proapoptotic Caspase Adaptor Protein Monoclonal specifically detects Proapoptotic Caspase Adaptor Protein in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Proapoptotic Caspase Adaptor Protein | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 22 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| caspase-2 binding protein, FLJ32987, HSPC190, marginal zone B and B1 cell-specific protein, MGC29506, PACAP, pERp1, plasma cell-induced ER protein 1, plasma cell-induced resident endoplasmic reticulum protein, Plasma cell-induced resident ER protein, Proapoptotic caspase adapter protein, proapoptotic caspase adaptor protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Proapoptotic Caspase Adaptor Protein (Q8WU39-1; Met 1- Thr 185). | |
| 0.1 mL | |
| Apoptosis, Caspases | |
| 51237 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction