missing translation for 'onlineSavingsMsg'
Learn More

Proadifen hydrochloride, MedChemExpress

Product Code. 16478726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478726 100 mg 100mg
16442266 500 mg 500mg
16429885 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478726 Supplier Medchem Express Supplier No. HYB1311
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Proadifen hydrochloride is a Cytochrome P450 inhibitor (IC50 = 19μM).

Proadifen hydrochloride is a Cytochrome P450 inhibitor (IC50 = 19μM).

Specifications

Chemical Name or Material Proadifen hydrochloride
CAS 62-68-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H32ClNO2
Quantity 100 mg
Synonym SKF-525A U-5446 RP-5171
Solubility Information DMSO : 50 mg/mL (128.22 mM; Need ultrasonic)
SMILES O=C(OCCN(CC)CC)C(CCC)(C1=CC=CC=C1)C2=CC=CC=C2.[H]Cl
Molecular Weight (g/mol) 389.96
Formula Weight 389.96
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.