missing translation for 'onlineSavingsMsg'
Learn More

Techsit™ Pro-Tech Laboratory High Chair

Product Code. 16593904
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Gray
With Armrest:
No
Yes
With Casters:
No
Yes
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Color With Armrest With Casters Förpackningsstorlek
16593904 Black Yes Yes Each
16503914 Black No Yes Each
16513914 Gray No No Each
16523914 Gray Yes No Each
16533914 Gray No Yes Each
16583904 Black No No Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16593904 Leverantör Techsit™ Leverantörsnummer TS7002B/AA
Estimated Shipment: 26-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Advanced laboratory high chair is fully adjustable with an adjustable foot ring.

  • Vinyl high chair with adjustable foot ring
  • Fully adjustable seat height, back height, and seat depth
  • Ergonomically designed chair that automatically adjusts to the user
  • Cleanroom Class 7 suitable

Specifikationer

Product Type Laboratory Seat
Color Black
Adjustable Yes
Arm Style Adjustable Arms
Autoclavable Not Autoclavable
Back Style Ergonomic
Finish Vinyl
Frame Material Steel
Lift Type Gas Lift
Load Bearing Capacity (Metric) 150 kg
Seating Height High Bench Height
Upholstery Vinyl
Warranty 5 Year
Weight (Metric) 17 kg
Wheel Type Glides
Width (Metric) 490 mm
With Armrest Yes
With Backrest Yes
With Casters Yes
Footrest Yes
With Lumbar Support Yes
With Tilt Yes
Material Vinyl
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.