Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Staining Kits
Description
The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. The PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.
Features include:
• Sensitive—detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis
• Built-in controls—molecular weight marker allows for approximation of protein size and serves as positive and negative control with unique mixture of phosphorylated and non-phosphorylated proteins
• Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitation
Specifications
Specifications
| Content And Storage | • Pro-Q™ Diamond Phosphoprotein Gel Stain, 200 mL •PeppermintStick™ Phosphoprotein Molecular Weight Standard, 40 μL |
| Description | Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 200 mL |
| Detection Location | In-Gel Detection |
| Detection Method | Fluorescence |
| Label or Dye | Pro-Q Diamond |
| Quantity | 200 mL Stain, 40 μL Standard Kit |
| Target Molecule | Proteins (Phosphoproteins) |
| Product Line | PRO-Q |
| Product Type | Phosphoprotein Gel Staining Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.