Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- 5 day turn around time
Specifications
Specifications
| For Use With (Equipment) | 8-channel Pipettes |
| Certifications/Compliance | ISO 17025 Accredited Service |
| Description | Data collected before and after repair |
| Calibration Points | 5 x 3 Measurements |
| Duration | 5 Days |
| Product Type | Pipette Pro PM and Calibration Service |
| Service Offering | As-found/After Repair Calibration |