missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pro-Detect™ Rapid Rabbit Fc Assay Kit

Product Code. 15930513 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 assays
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15930513

Brand: Thermo Scientific™ A38505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

10 assays

Pro-Detect™ Rapid Protein Fc Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of human, rabbit or mouse Fc-tagged proteins in protein expression and purification systems.

  • Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the Fc tag to provide a visual, color readout for detection within ten minutes
  • Detect tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
  • Easy to adopt—lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Simple, fast method—simply dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  •  Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.
Detectable Analytes Rabbit Fc-tagged proteins
Assay Lateral Flow Rapid Assay
Detection Method Chromogenic
Product Line Pro-Detect
Product Type Assay
Specificity Antibody
Sufficient For 10 Assays
Target Species Rabbit
For Use With (Application) Lateral Flow Rapid Assay
Quantity 10 assays
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.