missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pro-Detect™ Rapid HA Competitive Assay Kit

Product Code. 15970513 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Assays
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15970513 10 Assays Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15970513 Supplier Thermo Scientific™ Supplier No. A38509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

10 assays

Pro-Detect™ Rapid Competitive Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of tagged proteins in protein expression and purification systems

  • Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a qualitative detection within ten minutes
  • Detect tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling
  • Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  • Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.
Detectable Analytes HA-tagged proteins
Assay Lateral Flow Rapid Assay
Detection Method Chromogenic
Product Line Pro-Detect
Product Type Assay
Specificity Protein tag
Sufficient For 10 Assays
Target Species Human
For Use With (Application) Lateral Flow Rapid Assay
Quantity 10 Assays
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.