missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pro-Detect™ Rapid DYKDDDDK Competitive Assay Kit
Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 Assays
Unit Size:
Pack of 10
Description
Includes
10 assays
- Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the tag to provide a qualitative detection within ten minutes
- Detect tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling
- Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
- Dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
- Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
- Specific—assay does not cross-react with common lysis and protein preparation reagents
- Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
- Long shelf life—stable for at least one year at 4°C
Specifications
Specifications
| Content And Storage | • 10 Rapid Assay Strips • Sample Diluent, 15 mL Sufficient for 10 assay determinations Store at 2–8°C. |
| Detectable Analytes | DYKDDDDK (FLAG)-tagged proteins |
| Assay | Lateral Flow Rapid Assay |
| Detection Method | Chromogenic |
| Product Line | Pro-Detect |
| Product Type | Assay |
| Specificity | Protein tag |
| Sufficient For | 10 Assays |
| For Use With (Application) | Lateral Flow Rapid Assay |
| Quantity | 10 Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction