missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRNPIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555524
This item is not returnable.
View return policy
Description
Immunogen sequence: FCSISTRKLA AHGFGASMAA MVSFPPQRYH YFLVLDFEAT CDKPQIHPQE IIEFPILKLN GRTMEIESTF HMYV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 99%.
Specifications
| PRNPIP | |
| Polyclonal | |
| Unconjugated | |
| ERI3 | |
| enhanced RNAi three prime mRNA exonuclease homolog 3; ERI1 exoribonuclease 3; ERI1 exoribonuclease family member 3; ERI3; exoribonuclease 3; H_RG158K20.1; I79_002990; PINT1; Prion interactor 1; prion protein-interacting protein; PRNPIP; PRNPIP1; WUGSC:H_RG158K20.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79033 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43414 | |
| ERI3 | |
| Recombinant protein corresponding to Human PRNPIP. Recombinant protein control fragment (Product #RP-94116). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction