missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRND Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26947-1-AP-150UL
This item is not returnable.
View return policy
Description
The PRND antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PRND. This antibody recognizes human, mouse, rat antigen. The PRND antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| PRND | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23627 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 350 μg/mL | |
| DOPPEL; DPL; MGC41841; PrPLP; dJ1068H6.4 | |
| Fusion Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 17-20, 30 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction