missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12557-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes all of isoforms of PRMT8.
Arginine methylation is a widespread posttranslational modification mediated by arginine methyltransferases, such as PRMT8. Arginine methylation is involved in a number of cellular processes, including DNA repair, RNA transcription, signal transduction, protein compartmentalization, and possibly protein translation.Specifications
| PRMT8 | |
| Polyclonal | |
| Unconjugated | |
| PRMT8 | |
| HRMT1L3, HRMT1L4, PRMT8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381813, 56341, 688502 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PAK3, Q9NR22 | |
| Prmt8 | |
| PRMT8 Fusion Protein Ag3166 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction