missing translation for 'onlineSavingsMsg'
Learn More

PRMT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18303967
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18303967 20 μg 20µL
18338391 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18303967 Supplier Novus Biologicals Supplier No. NBP31556820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRMT8 Polyclonal antibody specifically detects PRMT8 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT8
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Knockout Validated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias EC 2.1.1, EC 2.1.1.-, EC 2.1.1.77, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HMT1 hnRNP methyltransferase-like 4, HMT1 hnRNP methyltransferase-like 4 (S. cerevisiae), HRMT1L3, HRMT1L4, protein arginine methyltransferase 8, protein arginine N-methyltransferase 4, protein arginine N-methyltransferase 8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-380 of human PRMT8 (NP_062828.3). VIFARDKWLKPGGLMFPDRAALYVVAIEDRQYKDFKIHWWENVYGFDMTCIRDVAMKEPLVDIVDPKQVVTNACLIKEVDIYTVKTEELSFTSAFCLQIQRNDYVHALVTYFNIEFTKCHKKMGFSTAPDAPYTHWKQTVFYLEDYLTVRRGEEIYGTISMKPNAKNVRDLDFTVDLDFKG
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56341
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.