missing translation for 'onlineSavingsMsg'
Learn More

PRMT8 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30762042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762042 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762042 Supplier Novus Biologicals Supplier No. NBP346890IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRMT8 Polyclonal antibody specifically detects PRMT8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT8
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1, EC 2.1.1.-, EC 2.1.1.77, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HMT1 hnRNP methyltransferase-like 4, HMT1 hnRNP methyltransferase-like 4 (S. cerevisiae), HRMT1L3, HRMT1L4, protein arginine methyltransferase 8, protein arginine N-methyltransferase 4, protein arginine N-methyltransferase 8
Host Species Rabbit
Immunogen protein arginine methyltransferase 8. (Uniprot# Q9NR22)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56341
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.