missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT7 Antibody (PCRP-PRMT7-1A7) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14145JF635
This item is not returnable.
View return policy
Description
PRMT7 Monoclonal antibody specifically detects PRMT7 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| PRMT7 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| [Myelin basic protein]-arginine N-methyltransferase PRMT7, EC 2.1.1, EC 2.1.1.125, EC 2.1.1.126, FLJ10640, Histone-arginine N-methyltransferase PRMT7, KIAA1933EC 2.1.1.-, myelin basic protein-arginine N-methyltransferase, protein arginine methyltransferase 7, protein arginine N-methyltransferase 7 | |
| Recombinant full-length human PRMT7 protein (Uniprot: Q9NVM4) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-PRMT7-1A7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 54496 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction