missing translation for 'onlineSavingsMsg'
Learn More

PRMT7 Antibody (PCRP-PRMT7-1A7), DyLight 550, Novus Biologicals™

Product Code. 18329943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329943 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329943 Supplier Novus Biologicals Supplier No. NBP314145R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRMT7 Monoclonal antibody specifically detects PRMT7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT7
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-PRMT7-1A7
Conjugate DyLight 550
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias [Myelin basic protein]-arginine N-methyltransferase PRMT7, EC 2.1.1, EC 2.1.1.125, EC 2.1.1.126, FLJ10640, Histone-arginine N-methyltransferase PRMT7, KIAA1933EC 2.1.1.-, myelin basic protein-arginine N-methyltransferase, protein arginine methyltransferase 7, protein arginine N-methyltransferase 7
Host Species Mouse
Immunogen Recombinant full-length human PRMT7 protein (Uniprot: Q9NVM4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54496
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.