missing translation for 'onlineSavingsMsg'
Learn More

PRMT6 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18402269 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402269 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402269 Supplier Novus Biologicals Supplier No. NBP299773R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRMT6 Polyclonal antibody specifically detects PRMT6 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT6
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human PRMT6 (Accession#: NP_060607.2; Met 1-Asp 375)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.