Learn More
Invitrogen™ PRMT5 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C136)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548331
Description
Specificity: This antibody recognizes PRMT5 (Protein arginine N-methyltransferase 5). It binds to a folded domain, amino acids 269-637. PRMT5 is a arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA.
PRMT5 (protein arginine N-methyltransferase 5) is an arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. PRMT5 specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins Sm D1 and Sm D3; such methylation is required for the assembly and biogenesis of snRNP core particles.
Specifications
| PRMT5 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O14744 | |
| PRMT5 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| RAB-C136 | |
| Unconjugated | |
| PRMT5 | |
| 72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10419 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.