missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT4 Recombinant Rabbit Monoclonal Antibody (PSH05-08)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556391
This item is not returnable.
View return policy
Description
PRMT4 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in DNA packaging, transcription regulation, signal transduction, mRNA stability, metabolism of nascent pre-RNA, and transcriptional activation. Tissue specificity: Overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.
Specifications
| PRMT4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q4AE70, Q86X55, Q9WVG6 | |
| Carm1 | |
| Recombinant protein within human CARM1 aa 101-500/608. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PSH05-08 | |
| Unconjugated | |
| Carm1 | |
| Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10498, 363026, 59035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction