missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRMT4 Monoclonal Antibody (3H2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA515796

Product Code. 12897812

  • £486.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-15796 targets CARM1 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human, Non-human primate, Mouse and Rat samples. The MA5-15796 immunogen is purified recombinant fragment of human CARM1 expressed in E. Coli. MA5-15796 detects CARM1 which has a predicted molecular weight of approximately 65kDa.

Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in DNA packaging, transcription regulation, signal transduction, mRNA stability, metabolism of nascent pre-RNA, and transcriptional activation. Tissue specificity: Overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRMT4
Monoclonal
Conc. not determined
ascites with 0.03% sodium azide
Q4AE70, Q86X55, Q9WVG6
Carm1
Purified recombinant fragment of human CARM1 expressed in E. Coli.
RUO
10498, 363026, 59035
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
3H2
Unconjugated
Carm1
Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4
Mouse
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.