missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRMT3 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C372)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548337
This item is not returnable.
View return policy
Description
Specificity: This antibody recognizes PRMT3 (Protein arginine N-methyltransferase 3). It binds to a folded domain, amino acids 211-531. PRMT3 methylates (mono and asymmetric dimethylation) the guanidino nitrogens of arginyl residues in some proteins.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
Specifications
| PRMT3 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O60678 | |
| Prmt3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| RAB-C372 | |
| Unconjugated | |
| Prmt3 | |
| 2010005E20Rik; 2410018A17Rik; AL033309; heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Hrmt1l3; PRMT3; protein arginine methyltransferase 3; protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction