missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody (RAB-C372) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-20142-0.025mg
This item is not returnable.
View return policy
Description
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for ELISA, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
Specifications
| PRMT3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Human | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| RAB-C372 | |
| Unconjugated | |
| EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
| 0.025 mg | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 10196 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction