missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody (RAB-C372) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£190.00 - £494.00
Specifications
| Antigen | PRMT3 |
|---|---|
| Clone | RAB-C372 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618044
|
Novus Biologicals
NBP3-20143-0.2mg |
0.2 mg |
£494.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656354
|
Novus Biologicals
NBP3-20143-0.025mg |
0.025 mg |
£190.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| PRMT3 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| RAB-C372 | |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| PBS | |
| 10196 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title