missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRMT1 Recombinant Rabbit Monoclonal Antibody (K01_3L62)

Product Code. 30826325
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826325 100 μL
30828868 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826325 Supplier Invitrogen™ Supplier No. MA564400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: TGEEIFGTIG MRPN.

PRMT1 is a member of the protein arginine N-methyltransferase (PRMT) family. Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Increased expression of PRMT1 may play a role in various types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_3L62
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene PRMT1
Gene Accession No. Q63009, Q99873
Gene Alias 6720434D09Rik; ANM1; arginine N-methyltransferase 1; AW214366; HCP1; heterogeneous nuclear ribonucleoprotein methyltransferase 1-like 2; heterogeneous nuclear ribonucleoproteins methyltransferase-like 2; histone-arginine N-methyltransferase PRMT1; HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2; HMT1 hnRNP methyltransferase-like 2; HMT1 hnRNP methyltransferase-like 2 isoform 3; HMT2; Hrmt1l2; interferon receptor 1-bound protein 4; IR1B4; Mrmt1; PRMT1; protein arginine methyltransferase 1; protein arginine N-methyltransferase 1
Gene Symbols PRMT1
Host Species Rabbit
Immunogen A synthetic peptide of human PRMT1 (aa 327-340).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3276, 60421
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.