missing translation for 'onlineSavingsMsg'
Learn More

PRMT1 Rabbit anti-Human, Mouse, Rat, Clone: 10W10U4, Novus Biologicals™

Product Code. 18326946
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326946 20 μg 20µL
18302762 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18326946 Supplier Novus Biologicals Supplier No. NBP31642320UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PRMT1 Monoclonal antibody specifically detects PRMT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRMT1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10W10U4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PRMT1 (Q99873).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3276
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.