missing translation for 'onlineSavingsMsg'
Learn More

PRMT1 Antibody (S08-3H8), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen PRMT1
Clone S08-3H8
Concentration 0.3 mg/mL
Dilution Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18618083
View Documents
Novus Biologicals
NBP3-19745-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18679393
View Documents
Novus Biologicals
NBP3-19745-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRMT1 Monoclonal antibody specifically detects PRMT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PRMT1
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
3276
IgG
Affinity purified
S08-3H8
Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse
ANM1, EC 2.1.1, EC 2.1.1.-, EC 2.1.1.125, EC 2.1.1.77, HCP1, Histone-arginine N-methyltransferase PRMT1, HMT1 (hnRNP methyltransferase, S. cerevisiae)-like 2, HMT1 hnRNP methyltransferase-like 2 (S. cerevisiae), HMT2, HRMT1L2HMT1 hnRNP methyltransferase-like 2, Interferon receptor 1-bound protein 4, IR1B4protein arginine N-methyltransferase 1, protein arginine methyltransferase 1
A synthetic peptide of human PRMT1 (Uniprot # Q99873)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.