missing translation for 'onlineSavingsMsg'
Learn More

PRL/PTP4A Antibody (334407) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30738122 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738122 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738122 Supplier Novus Biologicals Supplier No. FAB32191JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRL/PTP4A Monoclonal antibody specifically detects PRL/PTP4A in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRL/PTP4A
Applications Immunohistochemistry
Classification Monoclonal
Clone 334407
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias HH72;PRL1;PRL-1;Protein tyrosine phosphatase type IVA 1;PTP(CAAX1);PTP4A1;PTPCAAX1
Host Species Mouse
Immunogen E. coli-derived recombinant human PRL-3, Ala2-Met173, Accession # O75365
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.