missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRL-2/PTP4A2 Rabbit anti-Human, Clone: 15, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90172-50UL
This item is not returnable.
View return policy
Description
PRL-2/PTP4A2 Monoclonal specifically detects PRL-2/PTP4A2 in Human samples. It is validated for ELISA.
Specifications
| PRL-2/PTP4A2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8073 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 15 | |
| ELISA 1:5000-1:10000 | |
| EC 3.1.3.48, HU-PP-1HH13, OV-1PTP4A, PRL-2HH7-2, PRL2protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA 2, protein tyrosine phosphatase type IVA, member 2, Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, PTP(CAAXII), PTPCAAX2phosphatase of regenerating liver 2, ptp-IV1a, ptp-IV1b | |
| Recombinant Human PRL-2/PTP4A2 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction