missing translation for 'onlineSavingsMsg'
Learn More

PRL-2/PTP4A2 Antibody (3C2), Novus Biologicals™

Product Code. 18376808 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376808 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376808 Supplier Novus Biologicals Supplier No. H00008073M07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRL-2/PTP4A2 Monoclonal antibody specifically detects PRL-2/PTP4A2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRL-2/PTP4A2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3C2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias EC 3.1.3.48, HU-PP-1HH13, OV-1PTP4A, PRL-2HH7-2, PRL2protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA 2, protein tyrosine phosphatase type IVA, member 2, Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, PTP(CAAXII), PTPCAAX2phosphatase of regenerating liver 2, ptp-IV1a, ptp-IV1b
Host Species Mouse
Immunogen PTP4A2 (NP_003470.1, 1 a.a. ~ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNRPAPVEISYENMRFLITHNPTNATLNKFTEELKKYGVTTLVRVCDATYDKAPVEKEGIHVLDWPFDDGAPPPNQIVDDWLNLLKTKFREEPGC
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8073
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.