missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRL-2/PTP4A2 Antibody (015), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90172AF350
This item is not returnable.
View return policy
Description
PRL-2/PTP4A2 Monoclonal specifically detects PRL-2/PTP4A2 in Human samples. It is validated for ELISA.
Specifications
| PRL-2/PTP4A2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8073 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 15 | |
| ELISA | |
| EC 3.1.3.48, HU-PP-1HH13, OV-1PTP4A, PRL-2HH7-2, PRL2protein tyrosine phosphatase IVA, protein tyrosine phosphatase IVA2, protein tyrosine phosphatase type IVA 2, protein tyrosine phosphatase type IVA, member 2, Protein-tyrosine phosphatase 4a2, Protein-tyrosine phosphatase of regenerating liver 2, PTP(CAAXII), PTPCAAX2phosphatase of regenerating liver 2, ptp-IV1a, ptp-IV1b | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PRL-2/PTP4A2 (Q12974-1; Asn2-Gln167). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction