missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536511
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 32 kDa. Purity is >95% by SDS-PAGE.
A group of proteins that are intimately involved in this process are the serine/ threonine (Ser/Thr) protein kinases. PRKY (protein kinase, Y-linked) and PRKX (protein kinase, X-linked) are members of the Ser/Thr protein kinase family, both of which belong to the subfamily of cAMP-dependent kinases. Encoded by a gene that is located near the pseudoautosomal region on chromosome Y, PRKY contains one protein kinase domain through which it catalyzes the ATP-dependent phosphorylation of target proteins. Functioning in a similar manner to PRXY, PRKX contains one protein kinase domain and is essential for macrophage differentiation and renal epithelial cell migration. Defects in the genes encoding PRKY and PRXX are associated with sex reversal disorder, namely XX in males and XY in females.
Specifications
| PRKY | |
| Polyclonal | |
| Unconjugated | |
| PRKY | |
| PRKXP3; PRKY; PRKYP; protein kinase, Y-linked, pseudogene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5616 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43930 | |
| PRKY | |
| Synthetic peptide corresponding to amino acids 61-110 of human PRKY. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction